Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Per2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011066.3 | NP_035196.2 | 12 - 13 | 81 | ||
GenBank mRNA | AF035830.1 | - | 12 - 13 | 81 | ||
AF036893.1 | - | 12 - 13 | 81 | |||
AK044658.1 | - | 12 - 13 | 81 | |||
AK122253.1 | - | 12 - 13 | 81 | |||
AK165556.1 | - | 12 - 13 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |