Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pygm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011224.1 | NP_035354.1 | 12 - 13 | 1569 | 82 | |
GenBank mRNA | AF124787.1 | - | 12 - 13 | 1569 | 82 | |
BC012961.1 | - | 12 - 13 | 1575 | 3989941 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |