Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011789.2 | NP_035919.2 | 1 - 2 | 160 | 61 | |
GenBank mRNA | AK140637.1 | - | 1 - 2 | 1133 | 61 | |
KF481611.1 | - | 2 - 3 | 275 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Detect Genomic DNA Mus musculus Probe spans exons |