Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ash2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080793.2 | NP_001074262.1 | 12 - 13 | 1441 | 112 | |
NM_001286207.1 | NP_001273136.1 | 12 - 13 | 1414 | 112 | ||
NM_011791.3 | NP_035921.2 | 12 - 13 | 1531 | 112 | ||
GenBank mRNA | AB020983.2 | - | 12 - 13 | 1513 | 112 | |
AK087934.1 | - | 12 - 13 | 1506 | 112 | ||
AK132471.1 | - | 11 - 12 | 1437 | 112 | ||
AK145576.1 | - | 12 - 13 | 1406 | 112 | ||
AK145790.1 | - | 12 - 13 | 1418 | 112 | ||
AK145967.1 | - | 12 - 13 | 1385 | 112 | ||
AK146254.1 | - | 12 - 13 | 1443 | 112 | ||
AK146599.1 | - | 12 - 13 | 1415 | 112 | ||
AK146756.1 | - | 12 - 13 | 1406 | 112 | ||
AK146938.1 | - | 12 - 13 | 1508 | 112 | ||
BC012957.1 | - | 12 - 13 | 1518 | 4211433 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |