Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ash2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080793.2 | NP_001074262.1 | 13 - 14 | 1545 | 76 | |
NM_001286207.1 | NP_001273136.1 | 13 - 14 | 1518 | 76 | ||
NM_011791.3 | NP_035921.2 | 13 - 14 | 1635 | 76 | ||
GenBank mRNA | AB020983.2 | - | 13 - 14 | 1617 | 76 | |
AK087934.1 | - | 13 - 14 | 1610 | 76 | ||
AK132471.1 | - | 12 - 13 | 1541 | 76 | ||
AK145576.1 | - | 13 - 14 | 1510 | 76 | ||
AK145790.1 | - | 13 - 14 | 1522 | 76 | ||
AK145967.1 | - | 13 - 14 | 1489 | 76 | ||
AK146254.1 | - | 13 - 14 | 1547 | 76 | ||
AK146599.1 | - | 13 - 14 | 1519 | 76 | ||
AK146756.1 | - | 13 - 14 | 1510 | 76 | ||
AK146938.1 | - | 13 - 14 | 1612 | 76 | ||
AK212085.1 | - | 2 - 3 | 113 | 76 | ||
BC012957.1 | - | 13 - 14 | 1622 | 4211433 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |