Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs2st1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011828.3 | NP_035958.3 | 4 - 5 | 950 | 80 | |
GenBank mRNA | AF060178.1 | - | 4 - 5 | 628 | 80 | |
AF169243.1 | - | 4 - 5 | 617 | 80 | ||
AK088698.1 | - | 4 - 5 | 926 | 80 | ||
AK165112.1 | - | 4 - 5 | 916 | 80 | ||
BC025443.1 | - | 4 - 5 | 878 | 5068428 | 80 | |
BC059008.1 | - | 4 - 5 | 707 | 6849136 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |