Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Twf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011876.3 | NP_036006.1 | 2 - 3 | 256 | 106 | |
GenBank mRNA | AK002699.1 | - | 1 - 2 | 171 | 106 | |
AK003997.1 | - | 1 - 2 | 151 | 106 | ||
AK084026.1 | - | 2 - 3 | 256 | 106 | ||
AK087422.1 | - | 1 - 2 | 171 | 106 | ||
AK155204.1 | - | 2 - 3 | 239 | 106 | ||
AK157886.1 | - | 2 - 3 | 201 | 106 | ||
AK166900.1 | - | 2 - 3 | 224 | 106 | ||
AK170441.1 | - | 2 - 3 | 204 | 106 | ||
AY267189.1 | - | 2 - 3 | 104 | 106 | ||
BC003338.1 | - | 2 - 3 | 203 | 3257230 | 106 | |
Y17808.1 | - | 2 - 3 | 121 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |