Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013464.4 | NP_038492.1 | 3 - 4 | 716 | 61 | |
GenBank mRNA | AF405560.1 | - | 3 - 4 | 350 | 61 | |
AF405561.1 | - | 3 - 4 | 350 | 61 | ||
AF405562.1 | - | 3 - 4 | 350 | 61 | ||
AF405563.1 | - | 3 - 4 | 350 | 61 | ||
AF405565.1 | - | 3 - 4 | 350 | 61 | ||
AF405566.1 | - | 3 - 4 | 350 | 61 | ||
AF405567.1 | - | 3 - 4 | 350 | 61 | ||
AF405568.1 | - | 3 - 4 | 350 | 61 | ||
AF405570.1 | - | 3 - 4 | 350 | 61 | ||
AK153678.1 | - | 3 - 4 | 717 | 61 | ||
AK155380.1 | - | 3 - 4 | 768 | 61 | ||
AY662080.1 | - | 3 - 4 | 350 | 61 | ||
AY662083.1 | - | 3 - 4 | 350 | 61 | ||
AY662084.1 | - | 3 - 4 | 350 | 61 | ||
D38416.1 | - | 3 - 4 | 365 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |