Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015765.2 | NP_056580.2 | 3 - 4 | 346 | 111 | |
GenBank mRNA | AK031489.1 | - | 285 | 111 | ||
AK145937.1 | - | 3 - 4 | 300 | 111 | ||
AK153397.1 | - | 3 - 4 | 308 | 111 | ||
BC002056.1 | - | 3 - 4 | 274 | 3493119 | 111 | |
U08215.1 | - | 3 - 4 | 306 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |