Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015765.2 | NP_056580.2 | 7 - 8 | 699 | 78 | |
GenBank mRNA | AK031489.1 | - | 638 | 78 | ||
AK145937.1 | - | 7 - 8 | 653 | 78 | ||
AK153397.1 | - | 7 - 8 | 661 | 78 | ||
BC002056.1 | - | 7 - 8 | 627 | 3493119 | 78 | |
BU505607.1 | - | 4 - 5 | 350 | 78 | ||
U08215.1 | - | 7 - 8 | 659 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |