Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276284.1 | NP_001263213.1 | 4 - 5 | 788 | 74 | |
NM_016666.3 | NP_057875.1 | 5 - 6 | 887 | 74 | ||
GenBank mRNA | AK011609.1 | - | 4 - 5 | 661 | 74 | |
AK146390.1 | - | 4 - 5 | 790 | 74 | ||
BC075614.1 | - | 4 - 5 | 659 | 5710504 | 74 | |
BQ175443.1 | - | 2 - 3 | 74 | |||
U85489.1 | - | 5 - 6 | 856 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |