Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159289.1 | NP_001152761.1 | 2 - 3 | 722 | 152 | |
NM_001159290.1 | NP_001152762.1 | 2 - 3 | 377 | 152 | ||
NM_001242934.1 | NP_001229863.1 | 2 - 3 | 722 | 152 | ||
NM_016707.3 | NP_057916.1 | 2 - 3 | 722 | 152 | ||
GenBank mRNA | AA008362.1 | - | 1 - 2 | 176 | 151 | |
AF051525.1 | - | 2 - 3 | 660 | 152 | ||
AF169037.1 | - | 2 - 3 | 660 | 152 | ||
AF186018.1 | - | 2 - 3 | 603 | 152 | ||
AK004395.1 | - | 2 - 3 | 598 | 152 | ||
AK043677.1 | - | 2 - 3 | 521 | 152 | ||
AK045556.1 | - | 2 - 3 | 378 | 152 | ||
AK140949.1 | - | 2 - 3 | 667 | 152 | ||
BC051418.1 | - | 2 - 3 | 510 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |