Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdlim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016798.3 | NP_058078.1 | 1 - 2 | 190 | 64 | |
GenBank mRNA | AF002283.1 | - | 1 - 2 | 214 | 64 | |
AK016447.1 | - | 1 - 2 | 167 | 64 | ||
AK078421.1 | - | 1 - 2 | 190 | 64 | ||
AK141331.1 | - | 1 - 2 | 189 | 64 | ||
AK164054.1 | - | 1 - 2 | 189 | 64 | ||
BC070418.1 | - | 1 - 2 | 156 | 5714572 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |