Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdlim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016798.3 | NP_058078.1 | 3 - 4 | 420 | 66 | |
GenBank mRNA | AF002283.1 | - | 3 - 4 | 444 | 66 | |
AK016447.1 | - | 3 - 4 | 397 | 66 | ||
AK078421.1 | - | 3 - 4 | 420 | 66 | ||
BC070418.1 | - | 3 - 4 | 386 | 5714572 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |