Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199023.1 | NP_001185952.1 | 7 - 8 | 1855 | 75 | |
NM_001199024.1 | NP_001185953.1 | 7 - 8 | 1855 | 75 | ||
NM_001199025.1 | NP_001185954.1 | 8 - 9 | 1360 | 75 | ||
NM_016915.4 | NP_058611.1 | 7 - 8 | 1401 | 75 | ||
GenBank mRNA | AF259401.1 | - | 6 - 7 | 1135 | 75 | |
AK144495.1 | - | 8 - 9 | 1234 | 75 | ||
AK153333.1 | - | 7 - 8 | 1661 | 75 | ||
BC003487.1 | - | 7 - 8 | 1194 | 2811357 | 75 | |
BC049778.1 | - | 7 - 8 | 1581 | 75 | ||
BC052845.1 | - | 7 - 8 | 1226 | 30054573 | 75 | |
BC057209.1 | - | 7 - 8 | 1448 | 5329147 | 75 | |
DV647961.1 | - | 3 - 4 | 355 | 75 | ||
U88624.2 | - | 6 - 7 | 1077 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |