Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018830.1 | NP_061300.1 | 5 - 6 | 562 | 74 | |
GenBank mRNA | AB037111.1 | - | 6 - 7 | 847 | 74 | |
AB037181.1 | - | 5 - 6 | 562 | 74 | ||
AK034493.1 | - | 5 - 6 | 793 | 74 | ||
AK041937.1 | - | 5 - 6 | 751 | 74 | ||
AK046540.1 | - | 5 - 6 | 756 | 74 | ||
AK047692.1 | - | 5 - 6 | 750 | 74 | ||
AK080951.1 | - | 5 - 6 | 739 | 74 | ||
AK166100.1 | - | 5 - 6 | 810 | 74 | ||
BC137898.1 | - | 5 - 6 | 545 | 8860917 | 74 | |
BC145016.1 | - | 5 - 6 | 548 | 9053564 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |