Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019458.3 | NP_062331.2 | 1 - 2 | 265 | 72 | |
GenBank mRNA | AB041615.1 | - | 1 - 2 | 283 | 72 | |
AK017762.1 | - | 1 - 2 | 335 | 72 | ||
AK134857.1 | - | 1 - 2 | 278 | 72 | ||
AK148538.1 | - | 1 - 2 | 300 | 72 | ||
AK151665.1 | - | 1 - 2 | 312 | 72 | ||
AK152778.1 | - | 1 - 2 | 281 | 72 | ||
AK152836.1 | - | 1 - 2 | 312 | 72 | ||
BC010317.1 | - | 1 - 2 | 220 | 72 | ||
BC029843.1 | - | 1 - 2 | 265 | 2651081 | 72 | |
BC083337.1 | - | 1 - 2 | 246 | 5701517 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |