Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293648.1 | NP_001280577.1 | 13 - 14 | 2198 | 76 | |
NM_019676.3 | NP_062650.1 | 13 - 14 | 2149 | 76 | ||
NR_121571.1 | 13 - 14 | 2207 | 76 | |||
GenBank mRNA | AF133125.1 | - | 13 - 14 | 2131 | 76 | |
AK028749.1 | - | 13 - 14 | 2150 | 76 | ||
AK082890.1 | - | 13 - 14 | 2141 | 76 | ||
BC015249.1 | - | 13 - 14 | 2161 | 4241580 | 76 | |
BC025798.1 | - | 13 - 14 | 2125 | 5068610 | 76 | |
JN635596.1 | - | 13 - 14 | 2198 | 76 | ||
U85711.1 | - | 13 - 14 | 2143 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |