Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdgfc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019971.2 | NP_064355.1 | 1 - 2 | 1136 | 105 | |
GenBank mRNA | AF117608.1 | - | 1 - 2 | 312 | 105 | |
AF266467.1 | - | 1 - 2 | 1136 | 105 | ||
AF286725.1 | - | 1 - 2 | 115 | 105 | ||
AK033734.1 | - | 1 - 2 | 865 | 105 | ||
AK042767.1 | - | 1 - 2 | 883 | 105 | ||
AK052947.1 | - | 1 - 2 | 388 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |