Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021435.3 | NP_067410.1 | 6 - 7 | 686 | 61 | |
GenBank mRNA | AB041549.1 | - | 6 - 7 | 687 | 61 | |
AK029046.1 | - | 5 - 6 | 3307 | 61 | ||
AK029969.1 | - | 6 - 7 | 744 | 61 | ||
AK033913.1 | - | 6 - 7 | 616 | 61 | ||
AK042150.1 | - | 6 - 7 | 586 | 61 | ||
AK045791.1 | - | 6 - 7 | 629 | 61 | ||
AK138591.1 | - | 6 - 7 | 627 | 61 | ||
AK163299.1 | - | 1 - 2 | 154 | 61 | ||
BC016275.1 | - | 6 - 7 | 617 | 61 | ||
BC033512.1 | - | 6 - 7 | 611 | 4017336 | 61 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BG062065.1 | - | Chr. - - [-] on Build | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |