Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnkp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290764.1 | NP_001277693.1 | 9 - 10 | 1034 | 65 | |
NM_001290766.1 | NP_001277695.1 | 7 - 8 | 1018 | 65 | ||
NM_001290767.1 | NP_001277696.1 | 7 - 8 | 789 | 65 | ||
NM_021549.3 | NP_067524.2 | 8 - 9 | 1126 | 65 | ||
GenBank mRNA | AF129451.1 | - | 7 - 8 | 784 | 65 | |
AK049992.1 | - | 8 - 9 | 929 | 65 | ||
AK159535.1 | - | 8 - 9 | 892 | 65 | ||
AK159770.1 | - | 8 - 9 | 892 | 65 | ||
AK160020.1 | - | 8 - 9 | 892 | 65 | ||
AK160160.1 | - | 9 - 10 | 1031 | 65 | ||
AK172558.1 | - | 7 - 8 | 823 | 65 | ||
BC047269.1 | - | 6 - 7 | 1068 | 65 | ||
BC057659.1 | - | 8 - 9 | 928 | 2865792 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |