Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285800.1 | NP_001272729.1 | 3 - 4 | 363 | 65 | |
NM_022811.3 | NP_073722.1 | 3 - 4 | 363 | 65 | ||
GenBank mRNA | AK050780.1 | - | 311 | 65 | ||
AK083948.1 | - | 3 - 4 | 333 | 65 | ||
AK132556.1 | - | 3 - 4 | 334 | 65 | ||
BC034783.1 | - | 3 - 4 | 310 | 4022372 | 65 | |
D14336.1 | - | 3 - 4 | 326 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |