Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irak1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022986.4 | NP_075362.3 | 1 - 2 | 533 | 109 | |
GenBank mRNA | AF093135.1 | - | 1 - 2 | 316 | 109 | |
AK013036.2 | - | 1 - 2 | 338 | 109 | ||
AK020170.1 | - | 1 - 2 | 536 | 109 | ||
AK020448.1 | - | 1 - 2 | 308 | 109 | ||
AK045436.1 | - | 1 - 2 | 329 | 109 | ||
AK076594.1 | - | 1 - 2 | 341 | 109 | ||
BC046301.1 | - | 1 - 2 | 327 | 109 | ||
BC061099.1 | - | 1 - 2 | 320 | 6742251 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |