Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nuf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023284.3 | NP_075773.2 | 5 - 6 | 492 | 107 | |
GenBank mRNA | AF326732.1 | - | 6 - 7 | 533 | 107 | |
AK010351.1 | - | 5 - 6 | 414 | 107 | ||
AK132467.1 | - | 5 - 6 | 493 | 107 | ||
AK162249.1 | - | 6 - 7 | 528 | 107 | ||
BC020026.1 | - | 6 - 7 | 501 | 3587655 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |