Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024172.3 | NP_077134.1 | 2 - 3 | 374 | 59 | |
GenBank mRNA | AF338351.1 | - | 1 - 2 | 312 | 59 | |
AK013263.1 | - | 2 - 3 | 365 | 59 | ||
AK088469.1 | - | 2 - 3 | 350 | 59 | ||
BC014758.1 | - | 2 - 3 | 374 | 4014661 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |