Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025359.3 | NP_079635.1 | 1 - 2 | 233 | 61 | |
GenBank mRNA | AK008175.1 | - | 1 - 2 | 173 | 61 | |
AK012571.1 | - | 1 - 2 | 245 | 61 | ||
AK090217.1 | - | 1 - 2 | 245 | 61 | ||
AK090322.1 | - | 1 - 2 | 245 | 61 | ||
AK158761.1 | - | 1 - 2 | 238 | 61 | ||
AK165768.1 | - | 1 - 2 | 270 | 61 | ||
AK167495.1 | - | 1 - 2 | 285 | 61 | ||
AK167979.1 | - | 1 - 2 | 238 | 61 | ||
BC018317.1 | - | 1 - 2 | 233 | 4166625 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |