Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025417.2 | NP_079693.1 | 6 - 7 | 407 | 87 | |
GenBank mRNA | AK004174.1 | - | 6 - 7 | 408 | 87 | |
AK009802.1 | - | 6 - 7 | 406 | 87 | ||
AK172194.1 | - | 5 - 6 | 1519 | 87 | ||
BC033438.1 | - | 5 - 6 | 393 | 3995599 | 87 | |
BC049222.1 | - | 6 - 7 | 406 | 2649359 | 87 | |
BC055798.1 | - | 5 - 6 | 1498 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |