Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025446.3 | NP_079722.1 | 4 - 5 | 586 | 62 | |
GenBank mRNA | AK005332.1 | - | 4 - 5 | 614 | 62 | |
AK008210.1 | - | 4 - 5 | 589 | 62 | ||
AK162306.1 | - | 4 - 5 | 605 | 62 | ||
BC035305.1 | - | 4 - 5 | 736 | 3980923 | 62 | |
EF410113.1 | - | 2 - 3 | 471 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |