Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blzf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160208.1 | NP_001153680.1 | 6 - 7 | 1094 | 71 | |
NM_001160209.1 | NP_001153681.1 | 5 - 6 | 1181 | 71 | ||
NM_025505.4 | NP_079781.2 | 7 - 8 | 1115 | 71 | ||
GenBank mRNA | AK005489.1 | - | 5 - 6 | 915 | 71 | |
AK006544.1 | - | 6 - 7 | 1098 | 71 | ||
AK029787.1 | - | 7 - 8 | 1116 | 71 | ||
AK167493.1 | - | 5 - 6 | 1188 | 71 | ||
AK168933.1 | - | 5 - 6 | 942 | 71 | ||
AK171167.1 | - | 5 - 6 | 973 | 71 | ||
BC027025.1 | - | 5 - 6 | 956 | 5374749 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |