Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aptx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025444.3 | NP_001020615.1 | 1 - 2 | 56 | 70 | |
NM_001025445.2 | NP_001020616.1 | 1 - 2 | 56 | 70 | ||
GenBank mRNA | AK010516.1 | - | 1 - 2 | 43 | 70 | |
BB860888.1 | - | 1 - 2 | 60 | 70 | ||
BC021872.1 | - | 1 - 2 | 49 | 3994375 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |