Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mterf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025547.3 | NP_079823.2 | 6 - 7 | 1038 | 114 | |
GenBank mRNA | AK002299.1 | - | 6 - 7 | 979 | 114 | |
AK010525.1 | - | 6 - 7 | 1033 | 114 | ||
BC025173.1 | - | 6 - 7 | 1007 | 2655114 | 114 | |
BC085282.1 | - | 6 - 7 | 1008 | 30467998 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |