Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ska1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164355.1 | NP_001157827.1 | 5 - 6 | 689 | 164 | |
NM_025581.4 | NP_079857.3 | 5 - 6 | 753 | 164 | ||
GenBank mRNA | AK006442.1 | - | 5 - 6 | 650 | 164 | |
AK007138.1 | - | 4 - 5 | 521 | 164 | ||
AK013233.1 | - | 5 - 6 | 651 | 164 | ||
AK021226.1 | - | 5 - 6 | 653 | 164 | ||
AK172500.1 | - | 5 - 6 | 656 | 164 | ||
BC019940.1 | - | 5 - 6 | 604 | 3709649 | 164 | |
DQ926863.1 | - | 5 - 6 | 631 | 164 | ||
DQ926864.1 | - | 5 - 6 | 695 | 164 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |