Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080158.1 | NP_001073627.1 | 2 - 3 | 293 | 82 | |
NM_025639.4 | NP_079915.1 | 3 - 4 | 294 | 82 | ||
NM_178269.3 | NP_840000.2 | 3 - 4 | 294 | 82 | ||
GenBank mRNA | AK010885.1 | - | 3 - 4 | 286 | 82 | |
AK011191.1 | - | 3 - 4 | 271 | 82 | ||
AK011227.1 | - | 3 - 4 | 278 | 82 | ||
AK011464.1 | - | 3 - 4 | 295 | 82 | ||
AK011755.1 | - | 3 - 4 | 287 | 82 | ||
AK011793.1 | - | 3 - 4 | 280 | 82 | ||
AK017679.1 | - | 3 - 4 | 287 | 82 | ||
AK168203.1 | - | 3 - 4 | 275 | 82 | ||
AY243536.1 | - | 3 - 4 | 255 | 82 | ||
AY243537.1 | - | 3 - 4 | 255 | 82 | ||
BC009160.1 | - | 3 - 4 | 273 | 2655787 | 82 | |
BG276334.1 | - | 2 - 3 | 293 | 82 | ||
BY710734.1 | - | 3 - 4 | 295 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |