Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600005875.mm.1 |
TC0600000788.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600005875.mm.1 |
NM_001113564.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005875.mm.1 |
NM_001113564.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005875.mm.1 |
NM_001113565.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005875.mm.1 |
NM_001113565.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005875.mm.1 |
NM_001113566.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005875.mm.1 |
NM_001113566.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005875.mm.1 |
NM_025814.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005875.mm.1 |
NM_025814.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005879.mm.1 |
TC0600000788.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600005879.mm.1 |
NM_001113564.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005879.mm.1 |
NM_001113564.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005879.mm.1 |
NM_001113565.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005879.mm.1 |
NM_001113565.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005879.mm.1 |
NM_001113566.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005879.mm.1 |
NM_001113566.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005879.mm.1 |
NM_025814.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005879.mm.1 |
NM_025814.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005877.mm.1 |
TC0600000788.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600005877.mm.1 |
NM_001113564.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005877.mm.1 |
NM_001113564.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005877.mm.1 |
NM_001113565.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005877.mm.1 |
NM_001113565.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005877.mm.1 |
NM_001113566.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005877.mm.1 |
NM_001113566.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005877.mm.1 |
NM_025814.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005877.mm.1 |
NM_025814.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005878.mm.1 |
TC0600000788.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600005878.mm.1 |
NM_001113564.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005878.mm.1 |
NM_001113564.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005878.mm.1 |
NM_001113565.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005878.mm.1 |
NM_001113565.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005878.mm.1 |
NM_001113566.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005878.mm.1 |
NM_001113566.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005878.mm.1 |
NM_025814.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005878.mm.1 |
NM_025814.2 |
Refseq |