Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lman2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025828.3 | NP_080104.2 | 1 - 2 | 395 | 81 | |
GenBank mRNA | AK004952.1 | - | 1 - 2 | 226 | 81 | |
AK014384.1 | - | 1 - 2 | 217 | 81 | ||
AK028452.1 | - | 1 - 2 | 224 | 81 | ||
AK083442.1 | - | 1 - 2 | 227 | 81 | ||
AK129028.1 | - | 1 - 2 | 222 | 81 | ||
AK150383.1 | - | 1 - 2 | 224 | 81 | ||
AK166953.1 | - | 1 - 2 | 232 | 81 | ||
AK169401.1 | - | 1 - 2 | 223 | 81 | ||
BC049221.1 | - | 1 - 2 | 219 | 2599070 | 81 | |
BC055327.1 | - | 1 - 2 | 395 | 5702182 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |