Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acad8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025862.2 | NP_080138.2 | 2 - 3 | 257 | 81 | |
GenBank mRNA | AK009379.1 | - | 2 - 3 | 236 | 81 | |
AK019502.1 | - | 2 - 3 | 254 | 81 | ||
AK029897.1 | - | 2 - 3 | 221 | 81 | ||
AK051091.1 | - | 2 - 3 | 212 | 81 | ||
AK077335.1 | - | 2 - 3 | 237 | 81 | ||
AK161255.1 | - | 2 - 3 | 211 | 81 | ||
BC026535.1 | - | 2 - 3 | 231 | 81 | ||
BC037644.1 | - | 2 - 3 | 233 | 5053565 | 81 | |
BQ931973.1 | - | 2 - 3 | 257 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |