Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025867.2 | NP_080143.1 | 4 - 5 | 412 | 71 | |
GenBank mRNA | AK009003.1 | - | 4 - 5 | 414 | 71 | |
AK009855.1 | - | 4 - 5 | 413 | 71 | ||
AY367772.1 | - | 3 - 4 | 348 | 71 | ||
BC010313.1 | - | 3 - 4 | 379 | 2646766 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |