Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trappc6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025960.3 | NP_080236.2 | 3 - 4 | 290 | 100 | |
GenBank mRNA | AK007965.1 | - | 3 - 4 | 289 | 100 | |
AK008022.1 | - | 3 - 4 | 290 | 100 | ||
AK008462.2 | - | 3 - 4 | 289 | 100 | ||
AK088832.1 | - | 3 - 4 | 289 | 100 | ||
BC020128.1 | - | 2 - 3 | 239 | 100 | ||
BC037154.1 | - | 3 - 4 | 327 | 2811950 | 100 | |
BC106114.1 | - | 3 - 4 | 304 | 4168174 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |