Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkain1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025998.3 | NP_080274.2 | 1 - 2 | 208 | 72 | |
GenBank mRNA | AK011164.1 | - | 1 - 2 | 341 | 72 | |
AK011850.1 | - | 1 - 2 | 139 | 72 | ||
BC116391.1 | - | 1 - 2 | 138 | 40101865 | 72 | |
BC116392.1 | - | 1 - 2 | 138 | 72 | ||
CJ161499.1 | - | 1 - 2 | 208 | 72 | ||
EF058048.2 | - | 1 - 2 | 261 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |