Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301067.1 | NP_001287996.1 | 2 - 3 | 364 | 68 | |
NM_007598.4 | NP_031624.2 | 2 - 3 | 348 | 68 | ||
GenBank mRNA | AK053351.1 | - | 2 - 3 | 172 | 68 | |
AK128889.1 | - | 2 - 3 | 170 | 68 | ||
AK137222.1 | - | 2 - 3 | 153 | 68 | ||
AK144094.1 | - | 2 - 3 | 156 | 68 | ||
AK154002.1 | - | 2 - 3 | 368 | 68 | ||
AK156556.1 | - | 2 - 3 | 182 | 68 | ||
AK162629.1 | - | 2 - 3 | 153 | 68 | ||
AK165565.1 | - | 2 - 3 | 151 | 68 | ||
AK169652.1 | - | 2 - 3 | 143 | 68 | ||
AK170901.1 | - | 2 - 3 | 159 | 68 | ||
AK171388.1 | - | 2 - 3 | 166 | 68 | ||
BC005446.1 | - | 1 - 2 | 147 | 3600514 | 68 | |
BC005472.1 | - | 1 - 2 | 140 | 3590890 | 68 | |
L12367.1 | - | 2 - 3 | 174 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |