Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capza3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007605.4 | NP_031631.3 | 1 | 795 | 134 | |
GenBank mRNA | BC049620.1 | - | 1 | 763 | 6770504 | 134 |
D87471.1 | - | 1 | 753 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon |