Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpeb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252525.1 | NP_001239454.1 | 3 - 4 | 488 | 76 | |
NM_001252526.1 | NP_001239455.1 | 3 - 4 | 485 | 76 | ||
NM_007755.5 | NP_031781.1 | 3 - 4 | 485 | 76 | ||
GenBank mRNA | AK136088.1 | - | 3 - 4 | 488 | 76 | |
BC125476.1 | - | 3 - 4 | 405 | 40129991 | 76 | |
BC144948.1 | - | 3 - 4 | 402 | 76 | ||
BY720025.1 | - | 3 - 4 | 488 | 76 | ||
Y08260.1 | - | 3 - 4 | 404 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |