Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007757.2 | NP_031783.2 | 3 - 4 | 1000 | 121 | |
GenBank mRNA | AK157277.1 | - | 3 - 4 | 883 | 121 | |
AK166512.1 | - | 3 - 4 | 863 | 121 | ||
BC017680.1 | - | 3 - 4 | 857 | 4241687 | 121 | |
CA535096.1 | - | 3 - 4 | 328 | 121 | ||
D16333.1 | - | 3 - 4 | 914 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |