Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081437.1 | NP_001074906.1 | 12 - 13 | 2817 | 63 | |
NM_007992.2 | NP_032018.2 | 13 - 14 | 2958 | 63 | ||
GenBank mRNA | AK087922.1 | - | 1 - 2 | 203 | 63 | |
AK159361.1 | - | 12 - 13 | 3129 | 63 | ||
AK161522.1 | - | 12 - 13 | 2760 | 63 | ||
AK168688.1 | - | 12 - 13 | 2755 | 63 | ||
BC005443.1 | - | 12 - 13 | 2797 | 3490759 | 63 | |
X75285.1 | - | 13 - 14 | 2858 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |