Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166368.1 | NP_001159840.1 | 4 - 5 | 401 | 78 | |
NM_008020.3 | NP_032046.1 | 4 - 5 | 416 | 78 | ||
GenBank mRNA | AK165394.1 | - | 4 - 5 | 381 | 78 | |
BC031824.1 | - | 4 - 5 | 370 | 4196363 | 78 | |
BC053692.1 | - | 4 - 5 | 406 | 6395841 | 78 | |
CF949699.1 | - | 3 - 4 | 220 | 78 | ||
DV655383.1 | - | 4 - 5 | 401 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |