Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008065.2 | NP_032091.2 | 4 - 5 | 748 | 94 | |
GenBank mRNA | AK139916.1 | - | 4 - 5 | 428 | 94 | |
AK145446.1 | - | 4 - 5 | 428 | 94 | ||
AK145838.1 | - | 4 - 5 | 493 | 94 | ||
AK148087.1 | - | 4 - 5 | 519 | 94 | ||
AK150455.1 | - | 4 - 5 | 428 | 94 | ||
BC013562.1 | - | 2 - 3 | 143 | 3493987 | 94 | |
BC052448.1 | - | 4 - 5 | 748 | 6408997 | 94 | |
M74515.1 | - | 4 - 5 | 755 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |