Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008065.2 | NP_032091.2 | 8 - 9 | 1391 | 110 | |
GenBank mRNA | AK139916.1 | - | 8 - 9 | 1071 | 110 | |
AK145446.1 | - | 8 - 9 | 1071 | 110 | ||
AK145838.1 | - | 8 - 9 | 1136 | 110 | ||
AK148087.1 | - | 8 - 9 | 1162 | 110 | ||
AK150455.1 | - | 8 - 9 | 1071 | 110 | ||
AY282796.1 | - | 1 - 2 | 143 | 110 | ||
AY282797.1 | - | 1 - 2 | 143 | 110 | ||
AY282798.1 | - | 1 - 2 | 143 | 110 | ||
AY282799.1 | - | 1 - 2 | 143 | 110 | ||
BC013562.1 | - | 6 - 7 | 786 | 3493987 | 110 | |
BC052448.1 | - | 8 - 9 | 1391 | 6408997 | 110 | |
M74515.1 | - | 8 - 9 | 1398 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |