Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008081.3 | NP_032107.1 | 2 - 3 | 269 | 78 | |
GenBank mRNA | BC139165.1 | - | 2 - 3 | 259 | 8862187 | 78 |
BC139166.1 | - | 2 - 3 | 259 | 8862188 | 78 | |
L30104.1 | - | 2 - 3 | 269 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |