Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008081.3 | NP_032107.1 | 8 - 9 | 1008 | 62 | |
GenBank mRNA | BC071231.1 | - | 8 - 9 | 976 | 62 | |
BC139165.1 | - | 8 - 9 | 998 | 8862187 | 62 | |
BC139166.1 | - | 8 - 9 | 998 | 8862188 | 62 | |
L30104.1 | - | 8 - 9 | 1008 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |