Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK033196.1 | - | 4 - 5 | 483 | 77 | |
AK051795.1 | - | 4 - 5 | 453 | 77 | ||
AK133826.1 | - | 4 - 5 | 484 | 77 | ||
BC090645.1 | - | 4 - 5 | 500 | 77 | ||
BC094416.1 | - | 4 - 5 | 448 | 5707724 | 77 | |
CF948662.1 | - | 4 - 5 | 500 | 77 | ||
D26090.1 | - | 3 - 4 | 506 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |